1-(3,5-Dimethoxyphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183474-1G 183474-500MG 183474-5G
77466-214EA 594.31 USD
77466-214 77478-958 77489-192
1-(3,5-Dimethoxyphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%
1-(3,5-Dimethoxyphenyl)-2,2,3,3,3-pentafluoro-1-propanone

Precaución: For research use only

Formula: C₁₁H₉F₅O₃
MW: 284.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16615530
CAS Number: 121592-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR