2,2,3,3,3-Pentafluoro-1-(3-methylphenyl)-1-propanone ≥96%

Supplier: AOBChem USA

Warning

183471-1G 183471-500MG 183471-5G 183471-25G
77466-210EA 284.28 USD
77466-210 77478-954 77489-188 77499-602
2,2,3,3,3-Pentafluoro-1-(3-methylphenyl)-1-propanone ≥96%
2,2,3,3,3-Pentafluoro-1-(3-methylphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₀H₇F₅O
MW: 238.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16615531
CAS Number: 73741-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR