2,2,3,3,3-Pentafluoro-1-(4-fluorophenyl)-1-propanone ≥97%

Supplier: AOBChem USA

Warning

183467-1G 183467-500MG 183467-5G
77466-206EA 594.31 USD
77466-206 77478-950 77489-184
2,2,3,3,3-Pentafluoro-1-(4-fluorophenyl)-1-propanone ≥97%
2,2,3,3,3-Pentafluoro-1-(4-fluorophenyl)-1-propanone

Precaución: For research use only

Formula: C₉H₄F₆O
Storage Temperature: Ambient
MDL Number: MFCD16613960
CAS Number: 80180-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR