5-Bromo-1,3-difluoro-2-[tris(1-methylethyl)silyl]benzene ≥97%

Supplier: AOBChem USA

Warning

183435-1G 183435-500MG 183435-5G 183435-25G
77466-202EA 415.09 USD
77466-202 77478-946 77489-180 77499-598
5-Bromo-1,3-difluoro-2-[tris(1-methylethyl)silyl]benzene ≥97%
5-Bromo-1,3-difluoro-2-[tris(1-methylethyl)silyl]benzene

Precaución: For research use only

Formula: C₁₅H₂₃BrF₂Si
MW: 349.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30179086
CAS Number: 872545-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR