1-(5-Bromo-2-fluorophenyl)-2-cyclohexen-1-ol ≥95%

Supplier: AOBChem USA

Warning

183347-1G 183347-500MG 183347-5G
77466-198EA 597.4 USD
77466-198 77478-942 77489-176
1-(5-Bromo-2-fluorophenyl)-2-cyclohexen-1-ol ≥95%
1-(5-Bromo-2-fluorophenyl)-2-cyclohexen-1-ol

Precaución: For research use only

Formula: C₁₂H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD34857679
CAS Number: 1597255-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR