2,5-Dimethyl-α,α-bis(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

183234-1G 183234-500MG 183234-5G 183234-25G
77466-194EA 415.09 USD
77466-194 77478-938 77489-172 77499-592
2,5-Dimethyl-α,α-bis(1-methylethyl)benzenemethanol ≥95%
2,5-Dimethyl-α,α-bis(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857591
CAS Number: 1866292-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR