α,α-Bis(1-methylethyl)-3-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

183200-1G 183200-500MG 183200-5G 183200-25G
77466-190EA 415.09 USD
77466-190 77478-934 77489-168 77499-588
α,α-Bis(1-methylethyl)-3-(methylthio)benzenemethanol ≥95%
α,α-Bis(1-methylethyl)-3-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂OS
MW: 238.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857558
CAS Number: 2153124-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR