α-Methyl-3-(methylthio)-α-propylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

183122-1G 183122-500MG 183122-5G 183122-25G
77466-186EA 352.26 USD
77466-186 77478-930 77489-164 77499-584
α-Methyl-3-(methylthio)-α-propylbenzenemethanol ≥95%
α-Methyl-3-(methylthio)-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈OS
Storage Temperature: Ambient
MDL Number: MFCD20528612
CAS Number: 1378876-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR