α-Methyl-3-(1-methylethyl)-α-propylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

183093-1G 183093-500MG 183093-5G 183093-25G
77466-182EA 341.96 USD
77466-182 77478-926 77489-160 77499-582
α-Methyl-3-(1-methylethyl)-α-propylbenzenemethanol ≥95%
α-Methyl-3-(1-methylethyl)-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD20526642
CAS Number: 1443304-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR