2,6-Difluoro-α,3-Dimethyl-α-propylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

183048-1G 183048-500MG 183048-5G
77466-174EA 530.45 USD
77466-174 77478-918 77489-152
2,6-Difluoro-α,3-Dimethyl-α-propylbenzenemethanol ≥95%
2,6-Difluoro-α,3-Dimethyl-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD24310389
CAS Number: 1551510-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR