4-(4-Ethoxyphenyl)tetrahydro-2H-thiopyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

182948-1G 182948-500MG 182948-5G 182948-25G
77466-170EA 415.09 USD
77466-170 77478-914 77489-148 77499-578
4-(4-Ethoxyphenyl)tetrahydro-2H-thiopyran-4-ol ≥95%
4-(4-Ethoxyphenyl)tetrahydro-2H-thiopyran-4-ol

Precaución: For research use only

Formula: C₁₃H₁₈O₂S
MW: 238.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31693983
CAS Number: 2008299-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR