Tetrahydro-4-[4-(2-methylpropyl)phenyl]-2H-thiopyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

182884-1G 182884-500MG 182884-5G 182884-25G
77466-166EA 415.09 USD
77466-166 77478-910 77489-144 77499-576
Tetrahydro-4-[4-(2-methylpropyl)phenyl]-2H-thiopyran-4-ol ≥95%
Tetrahydro-4-[4-(2-methylpropyl)phenyl]-2H-thiopyran-4-ol

Precaución: For research use only

Formula: C₁₅H₂₂OS
MW: 250.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31693981
CAS Number: 2254577-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR