4-(2-Bromo-3-fluorophenoxy)aniline ≥97%

Supplier: AOBChem USA

Warning

182845-1G 182845-500MG 182845-5G 182845-25G
77466-162EA 255.44 USD
77466-162 77478-906 77489-140 77499-572
4-(2-Bromo-3-fluorophenoxy)aniline ≥97%
4-(2-Bromo-3-fluorophenoxy)aniline

Precaución: For research use only

Formula: C₁₂H₉BrFNO
MW: 282.1083 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857421
CAS Number: 2273520-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR