(8-Oxo-5,6,7,8-tetrahydronaphthalen-2-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (8-Oxo-6,7-dihydro-5H-naphthalen-2-yl)boronic acid

18262-1G 18262-500MG 18262-5G 18262-25G
77466-158EA 225.57 USD
77466-158 77478-902 77489-134 77499-566
(8-Oxo-5,6,7,8-tetrahydronaphthalen-2-yl)boronic acid ≥95%
(8-Oxo-5,6,7,8-tetrahydronaphthalen-2-yl)boronic acid

Precaución: For research use only

Formula: C₁₀H₁₁BO₃
Storage Temperature: Ambient
MDL Number: MFCD28122670
CAS Number: 1135871-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR