1-(3-Bromophenyl)-1-cyclopropylethanol ≥96%

Supplier: AOBChem USA

Warning

182442-1G 182442-500MG 182442-5G 182442-25G
77466-154EA 226.6 USD
77466-154 77478-898 77489-130 77499-558
1-(3-Bromophenyl)-1-cyclopropylethanol ≥96%
1-(3-Bromophenyl)-1-cyclopropylethanol

Precaución: For research use only

Formula: C₁₁H₁₃BrO
MW: 241.1243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21328247
CAS Number: 1487388-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR