(3-Bromo-2-fluorophenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

182415-1G 182415-500MG 182415-5G 182415-25G
77466-150EA 397.58 USD
77466-150 77478-894 77489-126 77499-554
(3-Bromo-2-fluorophenyl)(cyclohexyl)methanol ≥95%
(3-Bromo-2-fluorophenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₆BrFO
MW: 287.1679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857210
CAS Number: 1698633-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR