Cyclohexyl(2,6-difluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

182411-1G 182411-500MG 182411-5G
77466-146EA 435.69 USD
77466-146 77478-890 77489-122
Cyclohexyl(2,6-difluorophenyl)methanol ≥97%
Cyclohexyl(2,6-difluorophenyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₆F₂O
MW: 226.2623 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16293513
CAS Number: 1340382-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR