Cyclohexyl(2-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

182403-1G 182403-500MG 182403-5G
77466-142EA 435.69 USD
77466-142 77478-886 77489-118
Cyclohexyl(2-fluorophenyl)methanol ≥97%
Cyclohexyl(2-fluorophenyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₇FO
MW: 208.2718 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16293488
CAS Number: 1087394-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR