1-(3-Bromo-2,6-difluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥96%

Supplier: AOBChem USA

Warning

182395-1G 182395-500MG 182395-5G 182395-25G
77466-138EA 415.09 USD
77466-138 77478-882 77489-114 77499-550
1-(3-Bromo-2,6-difluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥96%
1-(3-Bromo-2,6-difluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₉H₂BrF₇O
Storage Temperature: Ambient
MDL Number: MFCD34857209
CAS Number: 1986411-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR