1-(3-Bromo-2-fluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥96%

Supplier: AOBChem USA

Warning

182391-1G 182391-500MG 182391-5G 182391-25G
77466-134EA 415.09 USD
77466-134 77478-878 77489-110 77499-548
1-(3-Bromo-2-fluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥96%
1-(3-Bromo-2-fluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₉H₃BrF₆O
Storage Temperature: Ambient
MDL Number: MFCD34857208
CAS Number: 1930977-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR