2,2,3,3,3-pentafluoro-1-(4-methoxyphenyl)propan-1-one ≥97%

Supplier: AOBChem USA

Warning

182379-1G 182379-500MG 182379-5G 182379-25G
77466-126EA 373.89 USD
77466-126 77478-870 77489-102 77499-546
2,2,3,3,3-pentafluoro-1-(4-methoxyphenyl)propan-1-one ≥97%
2,2,3,3,3-pentafluoro-1-(4-methoxyphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₀H₇F₅O₂
Storage Temperature: Ambient
MDL Number: MFCD05864392
CAS Number: 345901-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR