2-(3-Bromo-2-fluorophenyl)pentan-2-ol ≥96%

Supplier: AOBChem USA

Warning

182371-1G 182371-500MG 182371-5G
77466-122EA 447.02 USD
77466-122 77478-866 77489-096
2-(3-Bromo-2-fluorophenyl)pentan-2-ol ≥96%
2-(3-Bromo-2-fluorophenyl)pentan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₄BrFO
MW: 261.1306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30197694
CAS Number: 1691878-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR