2-(3-Fluoro-2-methylphenyl)pentan-2-ol ≥96%

Supplier: AOBChem USA

Warning

182365-1G 182365-500MG 182365-5G
77466-118EA 648.9 USD
77466-118 77478-862 77489-092
2-(3-Fluoro-2-methylphenyl)pentan-2-ol ≥96%
2-(3-Fluoro-2-methylphenyl)pentan-2-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.2611 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528610
CAS Number: 1379354-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR