2-(4-Methoxyphenyl)pentan-2-ol ≥95%

Supplier: AOBChem USA

Warning

182357-1G 182357-500MG 182357-5G
77466-114EA 648.9 USD
77466-114 77478-858 77489-088
2-(4-Methoxyphenyl)pentan-2-ol ≥95%
2-(4-Methoxyphenyl)pentan-2-ol

Precaución: For research use only

Formula: C₁₂H₁₈O₂
MW: 194.2701 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06654207
CAS Number: 91763-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR