3-(4-Bromophenyl)-2,4-dimethylpentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

182348-1G 182348-500MG 182348-5G 182348-25G
77466-106EA 226.6 USD
77466-106 77478-850 77489-078 77499-532
3-(4-Bromophenyl)-2,4-dimethylpentan-3-ol ≥97%
3-(4-Bromophenyl)-2,4-dimethylpentan-3-ol

Precaución: For research use only

Formula: C₁₃H₁₉BrO
MW: 271.1933 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857201
CAS Number: 104417-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR