3-(3-Bromophenyl)-2,4-dimethylpentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

182346-1G 182346-500MG 182346-5G 182346-25G
77466-102EA 397.58 USD
77466-102 77478-846 77489-074 77499-528
3-(3-Bromophenyl)-2,4-dimethylpentan-3-ol ≥97%
3-(3-Bromophenyl)-2,4-dimethylpentan-3-ol

Precaución: For research use only

Formula: C₁₃H₁₉BrO
MW: 271.1933 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857199
CAS Number: 2012839-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR