2,4-Dimethyl-3-phenylpentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

182339-1G 182339-500MG 182339-5G 182339-25G
77466-098EA 297.67 USD
77466-098 77478-842 77489-070 77499-524
2,4-Dimethyl-3-phenylpentan-3-ol ≥97%
2,4-Dimethyl-3-phenylpentan-3-ol

Precaución: For research use only

Formula: C₁₃H₂₀O
MW: 192.2973 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857195
CAS Number: 4397-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR