2-Fluoro-3-methoxy-α,α,5-trimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

182302-1G 182302-500MG 182302-5G
77466-094EA 685.98 USD
77466-094 77478-838 77489-064
2-Fluoro-3-methoxy-α,α,5-trimethylbenzenemethanol ≥95%
2-Fluoro-3-methoxy-α,α,5-trimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₅FO₂
MW: 198.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28525266
CAS Number: 1783326-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR