2-Bromo-5-methylbenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

181652-1G 181652-500MG 181652-5G 181652-25G
77466-086EA 226.6 USD
77466-086 77478-830 77489-056 77499-510
2-Bromo-5-methylbenzo[b]thiophene ≥95%
2-Bromo-5-methylbenzo[b]thiophene

Precaución: For research use only

Formula: C₉H₇BrS
Storage Temperature: Ambient
MDL Number: MFCD05865140
CAS Number: 111860-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR