2-(Methylthio)dibenzofuran ≥95%

Supplier: AOBChem USA

Warning

181629-1G 181629-500MG 181629-5G 181629-25G
77466-082EA 415.09 USD
77466-082 77478-826 77489-052 77499-506
2-(Methylthio)dibenzofuran ≥95%
2-(Methylthio)dibenzofuran

Precaución: For research use only

Formula: C₁₃H₁₀OS
Storage Temperature: Ambient
MDL Number: MFCD34856975
CAS Number: 66623-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR