4-Fluoro-2-(methylsulfonyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

18157-1G 18157-500MG 18157-5G 18157-25G
77466-078EA 367.71 USD
77466-078 77478-822 77489-046 77499-498
4-Fluoro-2-(methylsulfonyl)phenylboronic acid ≥95%
4-Fluoro-2-(methylsulfonyl)phenylboronic acid

Precaución: For research use only

Formula: C₇H₈BFO₄S
MW: 218.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18311843
CAS Number: 2377607-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR