2,2-Difluoro-α-methyl-1,3-benzodioxole-4-methanol ≥95%

Supplier: AOBChem USA

Warning

181431-1G 181431-500MG 181431-5G
77466-074EA 434.66 USD
77466-074 77478-818 77489-042
2,2-Difluoro-α-methyl-1,3-benzodioxole-4-methanol ≥95%
2,2-Difluoro-α-methyl-1,3-benzodioxole-4-methanol

Precaución: For research use only

Formula: C₉H₈F₂O₃
MW: 202.16 g/mol
Storage Temperature: Ambient
CAS Number: 1894496-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR