3,4-Difluoro-α-propylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

181197-1G 181197-500MG 181197-5G 181197-25G
77466-066EA 339.9 USD
77466-066 77478-810 77489-034 77499-492
3,4-Difluoro-α-propylbenzenemethanol ≥96%
3,4-Difluoro-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₂F₂O
MW: 186.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521079
CAS Number: 1179299-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR