4-Chloro-2-fluoro-3-methyl-α-propylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

181058-1G 181058-500MG 181058-5G
77466-054EA 447.02 USD
77466-054 77478-798 77489-022
4-Chloro-2-fluoro-3-methyl-α-propylbenzenemethanol ≥95%
4-Chloro-2-fluoro-3-methyl-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄ClFO
MW: 216.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34856761
CAS Number: 1864550-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR