4-Ethyl-α-propylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

181029-1G 181029-500MG 181029-5G 181029-25G
77466-050EA 335.78 USD
77466-050 77478-794 77489-018 77499-484
4-Ethyl-α-propylbenzenemethanol ≥95%
4-Ethyl-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD11521094
CAS Number: 166337-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR