2,5-Dimethyl-α-propylbenzenemethanol ≥95%
Supplier: AOBChem USA
|
Warning
|
180977-1G
180977-500MG
180977-5G
77466-046EA
471.74
USD
77466-046
77478-790
77489-014
2,5-Dimethyl-α-propylbenzenemethanol ≥95%
2,5-Dimethyl-α-propylbenzenemethanol
Precaución: For research use only
Formula:
C₁₂H₁₈O Storage Temperature: Ambient |
MDL Number:
MFCD11521112 CAS Number: 854179-21-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...