α,α-Diethyl-2,5-Dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

180946-1G 180946-500MG 180946-5G
77466-042EA 668.47 USD
77466-042 77478-786 77489-010
α,α-Diethyl-2,5-Dimethylbenzenemethanol ≥95%
α,α-Diethyl-2,5-Dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O
MW: 192.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16794465
CAS Number: 1249852-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR