α,α-Diethyl-4-(2-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

180910-1G 180910-500MG 180910-5G
77466-038EA 668.47 USD
77466-038 77478-782 77489-006
α,α-Diethyl-4-(2-methylpropyl)benzenemethanol ≥95%
α,α-Diethyl-4-(2-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526652
CAS Number: 1443302-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR