α,α-Diethyl-3-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

180896-1G 180896-500MG 180896-5G
77466-030EA 648.9 USD
77466-030 77478-774 77488-998
α,α-Diethyl-3-(1-methylethyl)benzenemethanol ≥95%
α,α-Diethyl-3-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
MW: 206.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526651
CAS Number: 1443355-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR