2-Chloro-α,α-diethyl-6-fluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

180881-1G 180881-500MG 180881-5G
77466-026EA 447.02 USD
77466-026 77478-770 77488-994
2-Chloro-α,α-diethyl-6-fluorobenzenemethanol ≥95%
2-Chloro-α,α-diethyl-6-fluorobenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄ClFO
Storage Temperature: Ambient
MDL Number: MFCD30276506
CAS Number: 1865080-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR