α,α-Diethyl-4-methoxy-3,5-Dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

180855-1G 180855-500MG 180855-5G 180855-25G
77466-022EA 341.96 USD
77466-022 77478-766 77488-990 77499-476
α,α-Diethyl-4-methoxy-3,5-Dimethylbenzenemethanol ≥95%
α,α-Diethyl-4-methoxy-3,5-Dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
MW: 222.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528631
CAS Number: 853404-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR