α,α-Diethyl-1-naphthalenemethanol ≥95%

Supplier: AOBChem USA

Warning

180826-1G 180826-500MG 180826-5G 180826-25G
77466-018EA 341.96 USD
77466-018 77478-762 77488-986 77499-474
α,α-Diethyl-1-naphthalenemethanol ≥95%
α,α-Diethyl-1-naphthalenemethanol

Precaución: For research use only

Formula: C₁₅H₁₈O
MW: 214.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222298
CAS Number: 3099-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR