3-butoxy-2-fluoro-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

180361-1G 180361-500MG 180361-5G 180361-25G
77466-014EA 415.09 USD
77466-014 77478-758 77488-980 77499-468
3-butoxy-2-fluoro-4-methoxybenzaldehyde ≥97%
3-butoxy-2-fluoro-4-methoxybenzaldehyde

Precaución: For research use only

Formula: C₁₂H₁₅FO₃
MW: 226.2441 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34856450
CAS Number: 2969475-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR