2-butoxy-1-fluoro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

180356-1G 180356-500MG 180356-5G 180356-25G
77466-010EA 415.09 USD
77466-010 77478-754 77488-976 77499-464
2-butoxy-1-fluoro-3-methoxybenzene ≥97%
2-butoxy-1-fluoro-3-methoxybenzene

Precaución: For research use only

Formula: C₁₁H₁₅FO₂
MW: 198.2340 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34856445
CAS Number: 2969430-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR