4-Benzyloxy-1-bromo-2-trifluoromethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

18019-1G 18019-500MG 18019-5G 18019-25G
77466-006EA 188.49 USD
77466-006 77478-750 77488-972 77499-462
4-Benzyloxy-1-bromo-2-trifluoromethoxybenzene ≥95%
4-Benzyloxy-1-bromo-2-trifluoromethoxybenzene

Precaución: For research use only

Formula: C₁₄H₁₀BrF₃O₂
MW: 347.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28806011
CAS Number: 647856-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR