3-Bromo-6-chloro-2-(trifluoromethyl)pyridine ≥96%

Supplier: AOBChem USA

Warning

179995-1G 179995-500MG 179995-5G 179995-25G
77466-002EA 116.39 USD
77466-002 77478-746 77488-966 77499-454
3-Bromo-6-chloro-2-(trifluoromethyl)pyridine ≥96%
3-Bromo-6-chloro-2-(trifluoromethyl)pyridine

Precaución: For research use only

Formula: C₆H₂BrClF₃N
MW: 260.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16609904
CAS Number: 1227563-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR