N-(2-Bromo-4,5-dimethylphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

17967-1G 17967-500MG 17967-5G 17967-25G
77465-998EA 311.06 USD
77465-998 77478-742 77488-962 77499-450
N-(2-Bromo-4,5-dimethylphenyl)acetamide ≥97%
N-(2-Bromo-4,5-dimethylphenyl)acetamide

Precaución: For research use only

Formula: C₁₀H₁₂BrNO
MW: 242.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03079789
CAS Number: 22364-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR