2-Bromo-α-methyl-4-(trifluoromethyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

179513-1G 179513-500MG 179513-5G 179513-25G
77465-994EA 369.77 USD
77465-994 77478-738 77488-958 77499-446
2-Bromo-α-methyl-4-(trifluoromethyl)benzenemethanol ≥97%
2-Bromo-α-methyl-4-(trifluoromethyl)benzenemethanol

Precaución: For research use only

Formula: C₉H₈BrF₃O
MW: 269.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19303756
CAS Number: 349-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR