2-[4-Bromo-3-(trifluoromethyl)phenyl]-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

17907-1G 17907-500MG 17907-5G 17907-25G
77465-990EA 219.39 USD
77465-990 77478-734 77488-954 77499-442
2-[4-Bromo-3-(trifluoromethyl)phenyl]-1,3-dioxolane ≥95%
2-[4-Bromo-3-(trifluoromethyl)phenyl]-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₈BrF₃O₂
MW: 297.069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619392
CAS Number: 2221812-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR