1-(2,3-Dimethylphenyl)-1-propanone ≥96%

Supplier: AOBChem USA

Warning

178741-1G 178741-500MG 178741-5G 178741-25G
77465-986EA 85.49 USD
77465-986 77478-730 77488-950 77499-438
1-(2,3-Dimethylphenyl)-1-propanone ≥96%
1-(2,3-Dimethylphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617671
CAS Number: 35028-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR